4-Aminobenzamidin Dihydrochlorid

Lieferant: BACHEM BIOCHEMICA

4031123.0005 4031123.0025
BACE4031123.0005EA 0 EUR
BACE4031123.0005 BACE4031123.0025
4-Aminobenzamidin Dihydrochlorid
4-Aminobenzamidin Dihydrochlorid
Formel: C₇H₁₁Cl₂N₃
MDL: MFCD00013001
CAS-Nummer: 2498-50-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR