5-Brom-4-chlor-3-indolylphosphat Dinatriumsalz (BCIP-Na)

Lieferant: BACHEM BIOCHEMICA

Synonyme: 5-Bromo-4-chloro-1H-indol-3-yl phosphate sodium, X-phosphate disodium salt, Sodium 5-bromo-4-chloro-1H-indol-3-yl phosphate

4031125.0250
BACE4031125.0250EA 0 EUR
BACE4031125.0250
5-Brom-4-chlor-3-indolylphosphat Dinatriumsalz (BCIP-Na)
5-Brom-4-chlor-3-indolylphosphat Dinatriumsalz (BCIP-Na)
Formel: C₈H₄BrClNNa₂O₄P
Molecular Weight: 370.43 g/mol
MDL: MFCD00036757
CAS-Nummer: 102185-33-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR