D-Threonin

Lieferant: BACHEM BIOCHEMICA

4031299.0025 4031299.0100
BACE4031299.0025EA 0 EUR
BACE4031299.0025 BACE4031299.0100
D-Threonin
D-Threonin
Formel: C₄H₉NO₃
MDL: MFCD00064269
CAS-Nummer: 632-20-2
EINECS: 211-171-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR