DL-Valin

Lieferant: BACHEM BIOCHEMICA

Synonyme: (±)-2-Amino-3-methylbutansäure, H-DL-Val-OH, (±)-Valin, (±)-2-Aminoisovaleriansäure

4031331.0025
BACE4031331.0025EA 0 EUR
BACE4031331.0025
DL-Valin
DL-Valin
Formel: C₅H₁₁NO₂
Molecular Weight: 117.15 g/mol
Siedepunkt: 217
Schmelzpunkt: 86
Dichte: 1.352 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00004267
CAS-Nummer: 516-06-3
EINECS: 208-220-0
Merck Index: 12,10046

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR