D(-)-Penicillamin

Lieferant: BACHEM BIOCHEMICA

Synonyme: H-D-Pen-OH, (S)-2-Amino-3-mercapto-3-methylbutanoic acid, Penicillamine

4032629.0005
BACE4032629.0005EA 0 EUR
BACE4032629.0005
D(-)-Penicillamin
D(-)-Penicillamin
Formel: C₅H₁₁NO₂S
Molecular Weight: 149.2 g/mol
MDL: MFCD00064302
CAS-Nummer: 52-67-5
EINECS: 200-148-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR