1-Methylimidazol-2-carbonsäure
Lieferant: BACHEM BIOCHEMICA
4035264.0001
4035264.0005
BACE4035264.0001EA
0
EUR
BACE4035264.0001
BACE4035264.0005
1-Methylimidazol-2-carbonsäure
Formel:
C₅H₆N₂O₂ |
MDL:
MFCD01863421 CAS-Nummer: 20485-43-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...