(S)-1-Boc-piperidin-3-carbonsäure

Lieferant: BACHEM BIOCHEMICA

Synonyme: (S)-Boc-nipecotic acid, 1-Boc-L-nipecotic acid, (S)-1-(tert-Butoxycarbonyl)piperidine-3-carboxylic acid

4065479.0005
BACE4065479.0005EA 0 EUR
BACE4065479.0005
(S)-1-Boc-piperidin-3-carbonsäure
(S)-1-Boc-piperidin-3-carbonsäure
Formel: C₁₁H₁₉NO₄
Molecular Weight: 229.28 g/mol
MDL: MFCD02179172
CAS-Nummer: 88495-54-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR