L-Arginin Hydrochlorid, Multikompendial, J.T.Baker®
Lieferant: Avantor
Synonyme:
L(+)-α-Amino-δ-guanidinovaleriansäure Hydrochlorid, L(+)-Arginin Hydrochlorid, H-Arg-OH.HCl, (S)-(+)-α-Amino-δ-guanidinovaleriansäure Hydrochlorid, (S)-(+)-Arginin Hydrochlorid, H-L-Arg-OH.HCl
BAKR2051-06EA
0
EUR
BAKR2051-06
BAKR2051-07
BAKR2051-09
L-Arginin Hydrochlorid, Multikompendial, J.T.Baker®
L-Arginin Hydrochlorid
Bitte beachten Storage Condition - Preserve in well closed containers. Store in well-closed, light resistant containers.
Formel:
C₆H₁₅ClN₄O₂ Schmelzpunkt: 218 °C Dichte: 1,42 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00064550 CAS-Nummer: 1119-34-2 EINECS: 214-275-1 Merck Index: 13,00785 |
Spezifikation Testergebnisse
USP - Assay (C₆H₁₄N₄O₂·HCl) (dried basis) | 98.5 - 101.5 % |
USP - Identification | Passes test |
USP - Specific Rotation [Å]²º^D (+) | 21.4 - 23.6 ° |
USP - Loss on Drying at 105°C | ≤ 0.2 % |
USP - Residue on Ignition | ≤ 0.1 % |
USP - Sulfate (SO₄) | ≤ 0.03 % |
USP - Related Compounds - Individual Impurities | ≤ 0.5 % |
USP - Related Compounds - Total Impurities | ≤ 2.0 % |
USP - Chloride (Cl) | 16.5 - 17.1 % |
EP/BP - Assay (C₆H₁₄N₄O₂·HCl) (dried basis) | 98.5 - 101.0 % |
EP/BP - Identification A | Passes test |
EP/BP - Identification B | Passes test |
EP/BP - Identification E | Passes test |
EP/BP - Appearance of Solution | Passes test |
EP/BP - Specific Rotation [Å]²º^D (+) | 21.0 - 23.5 ° |
EP/BP-Ninhydrin-Positive Substances-Each | ≤ 0.2 % |
EP/BP-Ninhydrin-Positive Substances-Total Impurities | ≤ 0.5 % |
EP/BP - Sulfate (SO₄) | ≤ 300 ppm |
EP/BP - Ammonium (NH₄) | ≤ 0.02 % |
EP/BP - Iron (Fe) | ≤ 10 ppm |
EP/BP - Loss on Drying | ≤ 0.5 % |
EP/BP - Ash (sulfated) | ≤ 0.1 % |
JP - Assay (C₆H₁₄N₄O₂·HCl) (dried basis) | 98.5 - 101.0 % |
JP - Identification | Passes test |
JP - Optical Rotation (+) | 21.5 - 23.5 ° |
JP - pH (1 in 10) | 4.7 - 6.2 |
JP - Clarity and Color of Solution | Passes test |
JP - Sulfate (SO₄) | ≤ 0.028 % |
JP - Ammonium (NH₄) | ≤ 0.02 % |
JP - Heavy Metals (as Pb) | ≤ 20 ppm |
JP - Arsenic (As) | ≤ 2 ppm |
JP - Related Substances | Passes test |
JP - Loss on Drying at 105°C | ≤ 0.20 % |
JP - Residue after Ignition | ≤ 0.10 % |
Endotoxin Concentration, IU/mg, | For Information Only |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...