L-Arginin Hydrochlorid, Multikompendial, J.T.Baker®

Lieferant: Avantor

Synonyme: L(+)-α-Amino-δ-guanidinovaleriansäure Hydrochlorid, L(+)-Arginin Hydrochlorid, H-Arg-OH.HCl, (S)-(+)-α-Amino-δ-guanidinovaleriansäure Hydrochlorid, (S)-(+)-Arginin Hydrochlorid, H-L-Arg-OH.HCl

BAKR2051-06EA 0 EUR
BAKR2051-06 BAKR2051-07 BAKR2051-09
L-Arginin Hydrochlorid, Multikompendial, J.T.Baker®
L-Arginin Hydrochlorid

Bitte beachten Storage Condition - Preserve in well closed containers. Store in well-closed, light resistant containers.

Formel: C₆H₁₅ClN₄O₂
Schmelzpunkt: 218 °C
Dichte: 1,42 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00064550
CAS-Nummer: 1119-34-2
EINECS: 214-275-1
Merck Index: 13,00785

Bestellen

Spezifikation Testergebnisse

USP - Assay (C₆H₁₄N₄O₂·HCl) (dried basis) 98.5 - 101.5 %
USP - Identification Passes test
USP - Specific Rotation [Å]²º^D (+) 21.4 - 23.6 °
USP - Loss on Drying at 105°C ≤ 0.2 %
USP - Residue on Ignition ≤ 0.1 %
USP - Sulfate (SO₄) ≤ 0.03 %
USP - Related Compounds - Individual Impurities ≤ 0.5 %
USP - Related Compounds - Total Impurities ≤ 2.0 %
USP - Chloride (Cl) 16.5 - 17.1 %
EP/BP - Assay (C₆H₁₄N₄O₂·HCl) (dried basis) 98.5 - 101.0 %
EP/BP - Identification A Passes test
EP/BP - Identification B Passes test
EP/BP - Identification E Passes test
EP/BP - Appearance of Solution Passes test
EP/BP - Specific Rotation [Å]²º^D (+) 21.0 - 23.5 °
EP/BP-Ninhydrin-Positive Substances-Each ≤ 0.2 %
EP/BP-Ninhydrin-Positive Substances-Total Impurities ≤ 0.5 %
EP/BP - Sulfate (SO₄) ≤ 300 ppm
EP/BP - Ammonium (NH₄) ≤ 0.02 %
EP/BP - Iron (Fe) ≤ 10 ppm
EP/BP - Loss on Drying ≤ 0.5 %
EP/BP - Ash (sulfated) ≤ 0.1 %
JP - Assay (C₆H₁₄N₄O₂·HCl) (dried basis) 98.5 - 101.0 %
JP - Identification Passes test
JP - Optical Rotation (+) 21.5 - 23.5 °
JP - pH (1 in 10) 4.7 - 6.2
JP - Clarity and Color of Solution Passes test
JP - Sulfate (SO₄) ≤ 0.028 %
JP - Ammonium (NH₄) ≤ 0.02 %
JP - Heavy Metals (as Pb) ≤ 20 ppm
JP - Arsenic (As) ≤ 2 ppm
JP - Related Substances Passes test
JP - Loss on Drying at 105°C ≤ 0.20 %
JP - Residue after Ignition ≤ 0.10 %
Endotoxin Concentration, IU/mg, For Information Only


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR