Ultrapure water system, Barnstead™ GenPure™
Lieferant: Thermo Fisher Scientific
50131952 50136146 50136151 50136152 50136153 50136169 50136170 50136171 50136172 50131922 50131948 50131950 50131217
BARN50131952EA 9410 EUR
BARN50131952 BARN50136146 BARN50136151 BARN50136152 BARN50136153 BARN50136169 BARN50136170 BARN50136171 BARN50136172 BARN50131922 BARN50131948 BARN50131950 BARN50131217
Ultrapure water system, Barnstead™ GenPure™
Wasseraufbereitungssysteme Reinstwassersysteme
Suitable for even the most demanding and sensitive applications, the family of Thermo Scientific™ Barnstead™ GenPure™ water purification systems deliver ultrapure 18,2 MΩ.cm water with consistent quality.
  • UV oxidation reduces organic levels to <5 ppb, and protects against microorganisms (only on systems containing a UV lamp (UV))
  • Ultrafiltration removes pyrogens from the feed water
  • Feed water monitoring alerts users to fluctuations in feed water quality
  • Real-time monitor of the amount of organic materials in the product water shown on the display
  • USP-
More Product Information