Water purification system, Barnstead™ Pacific™ TII
Lieferant: Thermo Fisher Scientific
50132132 50131982 50132123 50132124 50132125 50132131
BARN50132132EA 8480 EUR
BARN50132132 BARN50131982 BARN50132123 BARN50132124 BARN50132125 BARN50132131
Water purification system, Barnstead™ Pacific™ TII
Wasseraufbereitungssysteme Reinstwassersysteme
The Pacific TII water purification system converts tap water into Type 2 high purity water.
  • Optional UV lamp prevents biological contamination
  • Space-saving design permits benchtop installation or wall mounting with integrated mounting bracket
  • Water is dispensed directly from storage tank
  • Automatic return to 'operating' mode ensures recirculation during standstill periods
  • The high performance reverse osmosis membrane removes approximately 98% of inorganic ions and 99% of all dissolved organic substances as well as microorganisms and particles
  • The cartridge
More Product Information