Ultrapure water system, Barnstead™ GenPure™
Lieferant: Thermo Fisher Scientific
50131952 50136146 50136151 50136152 50136153 50136169 50136170 50136171 50136172 50131922 50131948 50131950 50131217
BARN50131952EA 9410 EUR
BARN50131952 BARN50136146 BARN50136151 BARN50136152 BARN50136153 BARN50136169 BARN50136170 BARN50136171 BARN50136172 BARN50131922 BARN50131948 BARN50131950 BARN50131217
Ultrapure water system, Barnstead™ GenPure™
Wasseraufbereitungssysteme Reinstwassersysteme
Suitable for even the most demanding and sensitive applications, the family of Thermo Scientific™ Barnstead™ GenPure™ water purification systems deliver ultrapure 18,2 MΩ.cm water with consistent quality.
  • UV oxidation reduces organic levels to <5 ppb, and protects against microorganisms (only on systems containing a UV lamp (UV))
  • Ultrafiltration removes pyrogens from the feed water
  • Feed water monitoring alerts users to fluctuations in feed water quality
  • Real-time monitor of the amount of organic materials in the product water shown on the display
  • USP-compliant conductivity measurement with temperature compensation can be switched on or off
  • Feed water source: Potable tap water, pretreated by reverse osmosis, ion exchange or distillation
  • RS-232 interface enables data printing and satisfies GLP guidelines
  • User-friendly, illuminated four-line alpha-numeric displays and tilting control panels for optimal view
Special feature of GenPure Pro system:
- Volumetric dispensing: Dispense at the push of a button; control from 0,01 to 65 L with an accuracy of <0,5%
Special features of GenPure xCAD Plus system:
- Volumetric dispensing: Dispense at the push of a button; control from 0,01 to 65 L with an accuracy of <0,5%
- GenPure xCAD Plus remote dispenser for full control of the system
- Under-the-bench mounting: Ideal solution when bench space is limited
Ideal for molecular biology, microbiology and analytical chemistry applications (e.g. cell and tissue culture, PCR, DNA sequencing, electrophoresis, analytical chemistry, HPLC, CG, CG-MS, ICP-MS, AA, TOC Measurements).
Lieferumfang: All systems include a wall bracket, ultrapure polisher cartridge, sterile 0,2 µm filter, pressure regulator, UV lamp and/or ultrafilter where applicable.