CryoPlus™ Storage Systems
Lieferant: Thermo Fisher Scientific
7405
BARN7405EA 29301 EUR
BARN7405
CryoPlus™ Storage Systems
Kryokonservierungssysteme
Increase valuable laboratory storage space with Thermo Scientific™ CryoPlus™ storage systems, which store up to 38500 vials (2,0 ml). The ideal combination of liquid nitrogen, storage reliability and microprocessor technology, the CryoPlus series LN₂ storage system provides precise and accurate control over all parameters without complicated programming.
  • 16 preset audible and visual alarm combinations provide maximum product protection
  • Automatic defogger incorporated into the manual fill index to improve visibility during retrieval
  • Liquid level sensor and fill port within vacuum space, for maximum storage and sensor protection
  • Front-mounted key lock and rear-mounted recessed power switch; control panel can also be locked
  • Temperature sleeve (included) provides colder temperatures and efficient vapor phase operation
Electrical: 200/230 V
Bestellinformation:
Optional racks, canisters, frames and risers allow you to create an organised inventory system that provides optimum storage in liquid or vapour phase. Contact your customer service representative for details.
Bitte beachten
Warranty varies by country, please contact us for details.