Magnetic Stirrers, RT basic
Lieferant: Thermo Fisher Scientific
88880008 88880010 88880012
BARN88880008EA 190 EUR
BARN88880008 BARN88880010 BARN88880012
Magnetic Stirrers, RT basic
Rührer Magnetrührer
The RT basic plug and play magnetic stirrers have a small foot print and are ideal for routine stirring applications.
  • Two non-slip silicone plate covers provided with all units (1 black, 1 white)
  • Strong magnetic coupling to ensure consistent stirring
  • Polypropylene top and body provides excellent chemical resistance
  • Low power consumption
  • Detachable plug