CryoExtra™ High-Efficiency Cryogenic Storage Systems with Battery Back-up
Lieferant: Thermo Fisher Scientific
CE8120BB CE8140BB CE8180BB CE8194BB
BARNCE8120BBEA 32700 EUR
BARNCE8120BB BARNCE8140BB BARNCE8180BB BARNCE8194BB
CryoExtra™ High-Efficiency Cryogenic Storage Systems with Battery Back-up
Dewars und Kryogefäße
Designed for both liquid and vapor phase storage, the CryoExtra™ Series features automated temperature monitoring, microprocessor-based LN2 level control, and a built-in battery back-up systems, providing peace-of-mind for valuable samples.
  • CryoExtra accepts both vertical and horizontal racks
  • User comfort features and convenient, built-in workspace
  • Designed for vapor-phase storage
  • Folding step for user comfort
  • Outstanding temperature uniformity
  • Large LED temperature display
  • Lockable lid for sample protection
  • Advanced temperature monitor and alarms
More Product Information