BSOCOES (Bis[2-(N-succinimidyl-oxycarbonyloxy)ethyl] sulphone)

Lieferant: G Bioscienes

BC01
BC01EA 535 EUR
BC01
BSOCOES (Bis[2-(N-succinimidyl-oxycarbonyloxy)ethyl] sulphone)
BSOCOES (Bis[2-(N-succinimidyl-oxycarbonyloxy)ethyl] sulphone)

(Bis(2-[Succinimidooxycarbonyloxy]ethyl) sulfone Homobifunctional Cross Linkers.

Formel: C₁₄H₁₆N₂O₁₂S
Molecular Weight: 436,35 g/mol
MDL: MFCD00047123
CAS-Nummer: 57683-72-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=74bef60154b431c30e32482f90c9752c24659290&vid=5717ef9242ec18da3ea26f7cc1dc6a5fdd52ef57&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Spacer Arm (Å) 13
Reactive Toward Primary Amines
Membrane Permeable X
Water Soluble No
Cleavable/ Reversible Base

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR