DPDPB (1,4-Di[3'-(2'-pyridyldithio)propionamido]butane)

Lieferant: G Bioscienes

BC03
BC03EA 751 EUR
BC03
DPDPB (1,4-Di[3'-(2'-pyridyldithio)propionamido]butane)
DPDPB (1,4-Di[3'-(2'-pyridyldithio)propionamido]butane)

1,4-Di-(3’-[2’pyridyldithio]-propionamido) butane Homobifunctional Cross Linker.

Formel: C₂₀H₂₆N₄O₂S₄
MDL: MFCD00153604
CAS-Nummer: 141647-62-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c7e2f9b8214f8781e8f92f6b05fa843d25ae77dc&vid=581f3f2f3a137bdda610161666a6eff449d68309&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Spacer Arm (Å) 19.9
Reactive Toward Sulfhydryls
Membrane Permeable not done
Water Soluble No
Cleavable/ Reversible Reducing Agents (Thiols)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR