DST (Disuccinimidyl tartrate)

Lieferant: G Bioscienes

BC05
BC05EA 668 EUR
BC05
DST (Disuccinimidyl tartrate)
DST (Disuccinimidyl tartrate)

Disuccinimidyl tartrate Homobifunctional Cross Linker.

Formel: C₁₂H₁₂N₂O₁₀
Molecular Weight: 344.231 g/mol
MDL: MFCD03788224
CAS-Nummer: 77658-91-4

Bestellen

Spezifikation Testergebnisse

Spacer Arm (Å) 6.4
Reactive Toward Primary Amines
Membrane Permeable Yes
Water Soluble No
Cleavable/ Reversible Oxidizing Agents (Periodate)

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR