Sulpho-DST (Sulphodisuccinimidyl tartrate)
Lieferant: G Bioscienes
Synonyme:
Sulfo DST
BC06
BC06EA
648
EUR
BC06
Sulpho-DST (Sulphodisuccinimidyl tartrate)
Sulpho-DST (Sulphodisuccinimidyl tartrate)
Sulfodisuccinimidyl tartrate Homobifunctional Cross Linker.
Formel:
C₁₂H₁₂N₂O₁₆S₂ Molecular Weight: 504,357 g/mol |
CAS-Nummer:
118674-04-7 |
Spezifikation Testergebnisse
Spacer Arm (Å) | 6.4 |
Reactive Toward | Primary Amines |
Membrane Permeable | 0 |
Water Soluble | 1 |
Cleavable/ Reversible | Oxidizing Agents (Periodate) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...