DTSSP (3,3'-Dithiobis(sulphosuccinimidyl propionate))

Lieferant: G Bioscienes

Synonyme: DTSSP

BC08
BC08EA 668 EUR
BC08
DTSSP (3,3'-Dithiobis(sulphosuccinimidyl propionate))
DTSSP (3,3'-Dithiobis(sulphosuccinimidyl propionate))

3,3’-Dithiobis(sulfosuccinimidyl propionate) Homobifunctional Cross Linker.

Formel: C₁₄H₁₆N₂O₁₄S₄
Molecular Weight: 564.542 g/mol
MDL: MFCD01861949
CAS-Nummer: 81069-02-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4956e583e6ad2e539b2b5e51c100b822ab1ee6c2&vid=4956e583e6ad2e539b2b5e51c100b822ab1ee6c2&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Spacer Arm (Å) 12
Reactive Toward Primary Amines
Membrane Permeable No
Water Soluble Yes
Cleavable/ Reversible Reducing Agents (Thiols)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR