EGS [Ethylene glycolbis(succinic acid-N-hydroxysuccinimide ester)]

Lieferant: G Bioscienes

Synonyme: Bis(2,5-dioxopyrrolidin-1-yl) O,O'-ethane-1,2-diyl disuccinate, EGS, Ethylene glycol disuccinate di(N-succinimidyl) ester, Ethylene glycol bis(succinimidyl succinate), Di(N-succinimidyl) ethylene glycol disuccinate

BC09
BC09EA 777 EUR
BC09
EGS [Ethylene glycolbis(succinic acid-N-hydroxysuccinimide ester)]
EGS [Ethylene glycolbis(succinic acid-N-hydroxysuccinimide ester)]

Ethylene glycol bis(succinimidyl succinate) Homobifunctional Cross Linker.


  • Ideal for receptor ligand crosslinking

Formel: C₁₈H₂₀N₂O₁₂
Molecular Weight: 456,36 g/mol
MDL: MFCD00049057
CAS-Nummer: 70539-42-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=22ce55e0c0806b52a130a128c309864463d713a6&vid=22ce55e0c0806b52a130a128c309864463d713a6&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Spacer Arm (Å) 16.1
Reactive Toward Primary Amines
Membrane Permeable X
Water Soluble No
Cleavable/ Reversible Hydroxylamine

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR