Sulpho-MBS (3-Maleimidobenzoyl-N-hydroxysulphosuccinimide ester sodium salt)

Lieferant: G Bioscienes
Achtung

Synonyme: Sulfo MBS

BC12
BC12EA 715 EUR
BC12
Sulpho-MBS (3-Maleimidobenzoyl-N-hydroxysulphosuccinimide ester sodium salt)
Sulpho-MBS (3-Maleimidobenzoyl-N-hydroxysulphosuccinimide ester sodium salt)

m-Maleimidobenzoyl-N-hydroxysulfosuccinimide esterHeterobifunctional Cross Linker


Bitte beachten For Research Use Only. Not for use in diagnostic procedures.

Formel: C₁₅H₉N₂O₉S·Na
Molecular Weight: 416.295 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD01318594
CAS-Nummer: 92921-25-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f93cde6433be4d22ba1280248284622784111712&vid=f93cde6433be4d22ba1280248284622784111712&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Spacer Arm (Å) 9.9
Reactive Toward Primary Amine + Sulfhydryl
Membrane Permeable No
Water Soluble Yes
Cleavable/ Reversible No


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR