GMBS (N-Succinimidyl 4-maleimidobutyrate)

Lieferant: G Bioscienes

Synonyme: GMBS

BC13
BC13EA 668 EUR
BC13
GMBS (N-Succinimidyl 4-maleimidobutyrate)
GMBS (N-Succinimidyl 4-maleimidobutyrate)

N-γ-Maleimidobutyryloxysuccinimide ester Heterobifunctional Cross Linker

Formel: C₁₂H₁₂N₂O₆
Molecular Weight: 280,24 g/mol
MDL: MFCD00036817
CAS-Nummer: 80307-12-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=af04e2196185d10521870c059dc57226588502fd&vid=af04e2196185d10521870c059dc57226588502fd&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Spacer Arm (Å) 6.8
Reactive Toward Primary Amine + Sulfhydryl
Membrane Permeable X
Water Soluble No
Cleavable/ Reversible No

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR