6-Maleinimido-capronsäure-N-succinimidylester (EMCS)

Lieferant: G Bioscienes
Achtung

Synonyme: 2,5-Dioxopyrrolidin-1-yl-6-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)hexanoate

BC16
BC16EA 866 EUR
BC16
6-Maleinimido-capronsäure-N-succinimidylester (EMCS)
6-Maleinimido-capronsäure-N-succinimidylester (EMCS)

N-(ε-Maleimidocaproyloxy) succinimide ester Heterobifunctional Cross Linker.

Formel: C₁₄H₁₆N₂O₆
Molecular Weight: 308,29 g/mol
MDL: MFCD00043043
CAS-Nummer: 55750-63-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=347fb08d6c84fa4d08bcfb6a28af9c54f714da18&vid=347fb08d6c84fa4d08bcfb6a28af9c54f714da18&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Spacer Arm (Å) 9.4
Reactive Toward Primary Amine + Sulfhydryl
Membrane Permeable 1
Water Soluble 0
Cleavable/ Reversible 0

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR