Sulpho-EMCS (N-(ε-maleimidocaproyloxy) sulphosuccinimide ester)
Lieferant: G Bioscienes
|
Achtung
|
Synonyme:
Sulfo EMCS
BC17
BC17EA
668
EUR
BC17
Sulpho-EMCS (N-(ε-maleimidocaproyloxy) sulphosuccinimide ester)
Sulpho-EMCS (N-(ε-maleimidocaproyloxy) sulphosuccinimide ester)
N-(ε-Maleimidocaproyloxy) sulfo succinimide ester Heterobifunctional Cross Linker.
Spezifikation Testergebnisse
Spacer Arm (Å) | 9.4 |
Reactive Toward | Primary Amine + Sulfhydryl |
Membrane Permeable | 0 |
Water Soluble | 1 |
Cleavable/ Reversible | 0 |