Sulpho-EMCS (N-(ε-maleimidocaproyloxy) sulphosuccinimide ester)
Lieferant: G Bioscienes
Achtung
Synonyme: Sulfo EMCS
BC17
BC17EA 668 EUR
BC17
Sulpho-EMCS (N-(ε-maleimidocaproyloxy) sulphosuccinimide ester)
Sulpho-EMCS (N-(ε-maleimidocaproyloxy) sulphosuccinimide ester)
N-(ε-Maleimidocaproyloxy) sulfo succinimide ester Heterobifunctional Cross Linker.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1787e19574e4d9cd8a0bb2cf8e2d7f7707a0a0fb&vid=a9ff9327f86d9ef65a95f4d779fe21eeb62cbfe8&prid=web&clid=SAPDAM
Bestellen
Spezifikation Testergebnisse
Spacer Arm (Å) 9.4
Reactive Toward Primary Amine + Sulfhydryl
Membrane Permeable 0
Water Soluble 1
Cleavable/ Reversible 0