PMPI (4-Maleimidophenyl isocyanate)

Lieferant: G Bioscienes
Gefahr

Synonyme: PMPI

BC18
BC18EA 585 EUR
BC18
PMPI (4-Maleimidophenyl isocyanate)
PMPI (4-Maleimidophenyl isocyanate)

N-(ρ-Maleimidophenyl) isocyanate Heterobifunctional Cross Linkers.

Formel: C₁₁H₆N₂O₃
Molecular Weight: 214,177 g/mol
MDL: MFCD00270900
CAS-Nummer: 123457-83-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a3f69a5f45f00193396f2a34d246a2842820bfee&vid=a3f69a5f45f00193396f2a34d246a2842820bfee&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Spacer Arm (Å) 8.7
Reactive Toward Sulfhydryl + Hydroxyl
Membrane Permeable Not tested
Water Soluble No
Cleavable/ Reversible No

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR