SIAB (N-Succinimidyl (4-iodoacetyl)aminobenzoate)

Lieferant: G Bioscienes

Synonyme: SIAB

BC19
BC19EA 668 EUR
BC19
SIAB (N-Succinimidyl (4-iodoacetyl)aminobenzoate)
SIAB (N-Succinimidyl (4-iodoacetyl)aminobenzoate)

N-Succinimidyl(4-iodoacetyl)aminobenzoate Heterobifunctional Cross Linkers.


  • Ideal for enzyme-antibody crosslinking

Formel: C₁₃H₉IN₂O₅
Molecular Weight: 400.125 g/mol
MDL: MFCD00078537
CAS-Nummer: 72252-96-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a6200ea4975e6b34d89d6eb71c29ec4c826b9c49&vid=a6200ea4975e6b34d89d6eb71c29ec4c826b9c49&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Spacer Arm (Å) 10.6
Reactive Toward Primary Amine + Sulfhydryl
Membrane Permeable Yes
Water Soluble No
Cleavable/ Reversible No


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR