SMCC (N-Succinimidyl 4-(N-Maleimidomethyl)cyclohexanecarboxylate)

Lieferant: G Bioscienes

Synonyme: SMCC

BC20
BC20EA 484 EUR
BC20
SMCC (N-Succinimidyl 4-(N-Maleimidomethyl)cyclohexanecarboxylate)
SMCC (N-Succinimidyl 4-(N-Maleimidomethyl)cyclohexanecarboxylate)

Succinimidyl 4-(N-maleimidomethyl) cyclohexane-1-carboxylate Heterobifunctional Cross Linkers.


  • Ideal for enzyme-antibody crosslinking

Formel: C₁₆H₁₈N₂O₆
Molecular Weight: 334,33 g/mol
MDL: MFCD00009634
CAS-Nummer: 64987-85-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ac0e970dd04ffbd61768df20abfc5fd2eb8a267e&vid=ac0e970dd04ffbd61768df20abfc5fd2eb8a267e&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Spacer Arm (Å) 11.6
Reactive Toward Primary Amine + Sulfhydryl
Membrane Permeable X
Water Soluble No
Cleavable/ Reversible No

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR