Sulpho-SIAB (Sulphosuccinimidyl (4-iodoacetyl)aminobenzoate)
Lieferant: G Bioscienes
Synonyme:
Sulfo SIAB
BC22
BC22EA
760
EUR
BC22
Sulpho-SIAB (Sulphosuccinimidyl (4-iodoacetyl)aminobenzoate)
Sulpho-SIAB (Sulphosuccinimidyl (4-iodoacetyl)aminobenzoate)
N-Sulfosuccinimidyl(4-iodoacetyl)aminobenzoate Heterobifunctional Cross Linkers.
Spezifikation Testergebnisse
Spacer Arm (Å) | 10.6 |
Reactive Toward | Primary Amine + Sulfhydryl |
Membrane Permeable | No |
Water Soluble | X |
Cleavable/ Reversible | No |