Sulpho-SIAB (Sulphosuccinimidyl (4-iodoacetyl)aminobenzoate)
Lieferant: G Bioscienes
Synonyme: Sulfo SIAB
BC22
BC22EA 760 EUR
BC22
Sulpho-SIAB (Sulphosuccinimidyl (4-iodoacetyl)aminobenzoate)
Sulpho-SIAB (Sulphosuccinimidyl (4-iodoacetyl)aminobenzoate)
N-Sulfosuccinimidyl(4-iodoacetyl)aminobenzoate Heterobifunctional Cross Linkers.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=30544f2410427dc0c4ee6b64e7bfe9bb30a62adb&vid=19822c003f231135255229c7f6b6a8f8b8b23ac8&prid=web&clid=SAPDAM
Bestellen
Spezifikation Testergebnisse
Spacer Arm (Å) 10.6
Reactive Toward Primary Amine + Sulfhydryl
Membrane Permeable No
Water Soluble X
Cleavable/ Reversible No