Sulpho-SMCC (3-Sulpho-N-succinimidyl-4-(N-maleimidomethyl)cyclohexane-1-carboxylate sodium salt)
Lieferant: G Bioscienes
|
Achtung
|
Synonyme:
Sodium 1-((4-((2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)methyl)cyclohexane-1-carbonyl)oxy)-2,5-dioxopyrrolidine-3-sulphonate, Sulphosuccinimidyl-trans-4-(N-maleimidomethyl)cyclohexane-1-carboxylate, Sulpho-SMCC Sodium Salt, SMCC, Water-Soluble, 4-(N-Maleimidomethyl)cyclohexane-1-carboxylic acid 3-sulpho-N-hydroxysuccinimide ester sodium salt, sulpho-SMCC, Sulfo-SMCC
BC23
BC23EA
720
EUR
BC23
Sulpho-SMCC (3-Sulpho-N-succinimidyl-4-(N-maleimidomethyl)cyclohexane-1-carboxylate sodium salt)
Sulpho-SMCC (3-Sulpho-N-succinimidyl-4-(N-maleimidomethyl)cyclohexane-1-carboxylate sodium salt)
Ideal for enzyme-antibody crosslinking.
OneQuant™ Sulfo SMCC is also available: Single use vials to minimize waste. No weighing required
Formel:
C₁₆H₁₈N₂O₉S Molecular Weight: 436.37 g/mol |
MDL:
MFCD00054978 CAS-Nummer: 92921-24-9 |
|
Spezifikation Testergebnisse
Spacer Arm (Å) | 11.6 |
Reactive Toward | Primary Amine + Sulfhydryl |
Membrane Permeable | No |
Water Soluble | Yes |
Cleavable/ Reversible | No |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...