Sulpho-SMPB (Sulphosuccinimidyl 4-(4-maleimidophenyl)butyrate)
Lieferant: G Bioscienes
Synonyme:
Sulfo SMPB
BC24
BC24EA
716
EUR
BC24
Sulpho-SMPB (Sulphosuccinimidyl 4-(4-maleimidophenyl)butyrate)
Sulpho-SMPB (Sulphosuccinimidyl 4-(4-maleimidophenyl)butyrate)
Sulfo succinimidyl 4-(ρ-maleimidophenyl) butyrate
Formel:
C₁₈H₁₅N₂O₉S·Na Molecular Weight: 458,374 g/mol |
MDL:
MFCD00054981 CAS-Nummer: 92921-26-1 |
Spezifikation Testergebnisse
Spacer Arm (Å) | 14.5 |
Reactive Toward | Primary Amine + Sulfhydryl |
Membrane Permeable | No |
Water Soluble | X |
Cleavable/ Reversible | No |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...