Sulpho-SASD (Sulphosuccinimidyl 2-(4-azidosalicylamido)ethyl-1,3-dithiopropionate)
Lieferant: G Bioscienes
Synonyme:
Sulfo SASD
BC40
BC40EA
971
EUR
BC40
Sulpho-SASD (Sulphosuccinimidyl 2-(4-azidosalicylamido)ethyl-1,3-dithiopropionate)
Sulpho-SASD (Sulphosuccinimidyl 2-(4-azidosalicylamido)ethyl-1,3-dithiopropionate)
Sulfo SASD Sulfosuccinimidyl-2-(p-azidosalicylamido)ethyl-1,3-dithiopropionate
- Spacer Arm (Å): 18.9
- Reactive Toward: Primary Amines
- Membrane Permeable: NO
- Water Soluble: YES
- Cleavable/ Reversible: Reducing Agents (Thiol)
Formel:
C₁₆H₁₆N₅NaO₉S₃ Molecular Weight: 541,51 g/mol |
MDL:
MFCD01861963 CAS-Nummer: 144650-95-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...