2-Iminothiolane hydrochloride (Traut's Reagent)

Lieferant: G Bioscienes

BC95
BC95EA 668 EUR
BC95
2-Iminothiolane hydrochloride (Traut's Reagent)
2-Iminothiolane hydrochloride (Traut's Reagent)

Traut's Reagent is used for the preparation of disulfide bridges or for the generation of sulphhydryl groups for conjugation. This is a small, water-soluble compound that reacts with primary amines under mild conditions (pH 7 to 10, 25 °C) for thiolation (sulphhydryl addition).


Molecular weight: 137,63

Formel: C₄H₈ClNS
MDL: MFCD00039013
CAS-Nummer: 4781-83-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=37007353d509687f56afdce2f7f74b862fccdc2e&vid=102f4b1adfcb0fa4b52494442af6ad2b8e0324cf&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR