SATA (N-Succinimidyl S-Acetylthioglycolate)

Lieferant: G Bioscienes

Synonyme: N-Succinimidyl-S-acetylthioacetate, S-Acetylthioglycolic Acid N-Succinimidyl Ester, SATA, 2,5-Dioxopyrrolidin-1-yl 2-(acetylthio)acetate

BC96
BC96EA 395 EUR
BC96
SATA (N-Succinimidyl S-Acetylthioglycolate)
SATA (N-Succinimidyl S-Acetylthioglycolate)

SATA reacts with primary amines, adding protected sulphhydryl residues. It is soluble in DMSO and the sulphhydryl group can be used in coupling reactions.

Formel: C₈H₉NO₅S
Molecular Weight: 231,22 g/mol
MDL: MFCD00036891
CAS-Nummer: 76931-93-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=026a46327b20992d5008cea02f1a3820be6fe265&vid=9b6f22242d8bab5886b0cd5f4f8d2d214e0947ef&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR