Natrium-1-hydroxy-2,5-dioxo-3-pyrrolidinsulfonat

Lieferant: G Bioscienes

Synonyme: N-Hydroxy sulphosuccinimide sodium salt, Hydroxy-2,5-dioxopyrrolidine-3-sulphonic acid sodium salt, N-Hydroxysulphosuccinimide sodium salt, Sulfo NHS, Sulpho-N-Hydroxysuccinimide

BC97
BC97EA 535 EUR
BC97
Natrium-1-hydroxy-2,5-dioxo-3-pyrrolidinsulfonat
Sulpho-NHS (N-Hydroxysulphosuccinimide sodium salt)

Sulpho-NHS reacts primarily with primary amines to convert carboxyl groups to amine reactive sulfo-NHS esters. It is ideal for cross-linking, chemical labelling and solid support immobilisation. It is used to generate amine reactive esters from carboxylate groups and is primarily used to increase the efficiency of the carbodiimide EDC cross-linking. EDC reactions do not require Sulpho-NHS; however, its presence greatly enhances cross-linking efficiency and allows for a two-step process. EDC reacts with a carboxylate group to form an active ester, which is subject to rapid hydrolysis in aqueous buffers. The Sulfo-NHS increases the stability of the active intermediate and the resulting coupled product is identical to EDC reactions without sulpho-NHS.


Sulpho-NHS is water-soluble, long-lived, and hydrolyses relatively slowly in water and in aqueous buffer, unlike NHS.

Formel: C₄H₄NNaO₆S
Molecular Weight: 217.13 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00043100
CAS-Nummer: 106627-54-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=80a7fb7e22e339a440f4c855dc478eeaa42ab12a&vid=a9c03aa77ec2e7c953ff9bfd57dfd10e20afe4fc&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR