Sterile tubing welder, SCD® IIB
Lieferant: TERUMO BCT
3NC-C986
BCTE3NC-C986EA 45960 EUR
BCTE3NC-C986
Sterile tubing welder, SCD® IIB
Schweißzubehör
The SCD IIB welder creates strong, sterile tubing connections in about 30 seconds.
  • Compact and portable
  • Ideal for polyvinyl chloride (PVC), C-FLEX® tubing type
  • Simple and easy to use and requires no software
  • Create both wet-to-dry and dry-to-dry connections with a single welder
SCD IIB welder allows you to perform a sterile connection to transfer, sample or replace components, all while maintaining a functionally closed system in open labs and production areas. Designed for use with 5,5 mm outside diameter tubing with a wall thickness of 8 mm.