Blood collection needles, BD Vacutainer® Flashback
Lieferant: BD Medical
301746 301747 301747UK 301746UK
BDAM301746EA 0 EUR
BDAM301746 BDAM301747 BDAM301747UK BDAM301746UK
Blood collection needles, BD Vacutainer® Flashback
Injektionsnadeln
BD Vacutainer® Flashback blood collection needles are multi-sample needles that provide instant confirmation of needle placement, through a unique visual flashback indication.
  • Avoids the need to probe for a vein, and multiple sticks of the same patient in cases where the vein is hard to find