Blood collection tubes, EDTA, BD Vacutainer® with BD Hemogard™ closure
Lieferant: BD Medical
367525 368841 367836 367864 361017 367838UK 367525UK 368841UK 368860UK 368860 367862 367873 367862UK 368857UK 361017UK 368857 368499 367839UK 367839
BDAM367525EA 63.4 EUR
BDAM367525 BDAM368841 BDAM367836 BDAM367864 BDAM361017 BDAM367838UK BDAM367525UK BDAM368841UK BDAM368860UK BDAM368860 BDAM367862 BDAM367873 BDAM367862UK BDAM368857UK BDAM361017UK BDAM368857 BDAM368499 BDAM367839UK BDAM367839
Blood collection tubes, EDTA, BD Vacutainer® with BD Hemogard™ closure
Klinisches Diagnosezubehör Klinische Probenentnahme und Probenvorbereitung Entnahme und Vorbereitung von Blutproben
These EDTA tubes are used to obtain a whole blood or EDTA plasma sample.
  • K2 or K3 potassium salt of EDTA (Ethylene Diamine Tetra Acetic acid) is used as an anticoagulant
  • Aprotinin K3 EDTA tube is available for assays that require a proteolytic enzyme inhibitor
There are no functional differences between the two additives but physical difference between is K3EDTA is a liquid solution in the BD Vacutainer® glass tubes and K2 EDTA is a liquid solution that is spray coated onto the interior surface of the BD Vacutainer® plus tubes.
Lieferumfang: All tubes are supplied in boxes of 100/cases of 1000.