Rapid serum collection tube, RST, BD Vacutainer®
Lieferant: BD Medical
368774 368774UK
BDAM368774EA 342 EUR
BDAM368774 BDAM368774UK
Rapid serum collection tube, RST, BD Vacutainer®
Klinisches Diagnosezubehör Klinische Probenentnahme und Probenvorbereitung Entnahme und Vorbereitung von Blutproben
PET, clear.
  • Can be centrifuged 5 minutes after the blood sample is taken
  • The gel barrier optimises the sample workflow
  • Centrifuging conditions: 4000xg for 3 minutes at 23 to 27 ºC or 2000xg for 4 minutes at 23 to 27 ºC or 1500 to 2000xg for 10 minutes at 23 to 27 ºC
This tube combines the advantages of a thrombin based clot activator with a gel barrier and enables rapid results as well as optimising the process. The clot activator produces high quality serum.