BD Vacutainer® Eclipse™ Signal™ Blood Collection Needle
Lieferant: BD Medical
368835
BDAM368835EA 0 EUR
BDAM368835
BD Vacutainer® Eclipse™ Signal™ Blood Collection Needle
Injektionsnadeln
The BD Vacutainer® Eclipse™ Signal™ blood collection needles offers a clear dedicated window for in-vein confirmation with an extra thin-wall cannula for a fast blood collection experience.
The BD Eclipse™ Signal™ with the integrated holder offers an ergonomic shape and an integrated safety shield to support end user experience. Altogether, this can provide greater confidence during venous blood collection with ease of handling and increase healthcare worker safety.
Verpackung: 50 per box, 400 per case