BD® Blunt Fill NRFit™ Needles
Lieferant: BD Medical
400067
BDAM400067EA 385 EUR
BDAM400067
BD® Blunt Fill NRFit™ Needles
Injektionsnadeln
BD® offers NRFit™ blunt fill and blunt filter needles for your medication preparation needs.
  • BD® Regional anaesthesia NRFit™ neuraxial products are easily identifiable through every product and package having an NRFit™ label and a yellow component
  • With NRFit™ products from BD, the connection may be new, but the functionality and technique remain the same
  • The BD® blunt filter NRFit™ needle has a 5 µm filter to filter out particles such as, glass and plastic fragments, for safer medication delivery
  • Sterile, single use
  • Natural rubber latex is not part of the material formulation
The improved patient safety of NRFit™ connections combined with the quality, reliability and service of BD® allow you to manage the transition to a new standard of care with ease.