Procaspase-3 Aktivator (PAC1, (4-Benzylpiperazino)essigsäure-(3-allyl-2-hydroxybenzyliden)hydrazid)

Lieferant: BD
Achtung

Synonyme: 4-(Phenylmethyl)[[2-hydroxy-3-(2-propenyl)phenyl]methylene]hydrazide-1-piperazine acetic acid, (4-benzylpiperazino)acetic acid-(3-allyl-2-hydroxybenzylidene)hydrazide

340535
BDIM340535EA 1390 EUR
BDIM340535
Procaspase-3 Aktivator (PAC1, (4-Benzylpiperazino)essigsäure-(3-allyl-2-hydroxybenzyliden)hydrazid)
Procaspase-3 Aktivator (PAC1, (4-Benzylpiperazino)essigsäure-(3-allyl-2-hydroxybenzyliden)hydrazid)
Formel: C₂₃H₂₈N₄O₂
Molecular Weight: 392,49 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD03302441
CAS-Nummer: 315183-21-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR