Centrifuge Tube Slicer
Lieferant: Beckman Coulter
347960
BECM347960EA 0 EUR
BECM347960
Centrifuge Tube Slicer
Schneidewerkzeuge
This CentriTube slicer is used for slicing polypropylene, polycarbonate, and cellulose propionate centrifuge tubes with diameters of 5 to 13 mm.