J-Lite™ JLA-9.1000 Fixed-Angle Rotor
Lieferant: Beckman Coulter
C71484
BECMC71484EA 22860 EUR
BECMC71484
J-Lite™ JLA-9.1000 Fixed-Angle Rotor
Zentrifugen Rotoren
Lightweight, high-performance, four-place, aluminum, and fixed-angle rotors are for use in Avanti™ J series.
  • Ideal for general-purpose, large-volume applications such as pelleting of bacteria, subcellular organelles, viruses, and precipitates
  • Space for Four 1000 ml bottles
  • Lid with removable canisters
Material: Anodized aluminum