Bench top cell sorter, CytoFLEX SRT
Lieferant: Beckman Coulter
C71883 C71884 C71885 C71886 C71887 C71888 C71889 C71900 C71901 C71902 C71903 C71904 C71905 C71906 C85213 C85214
BECMC71883EA 0 EUR
BECMC71883 BECMC71884 BECMC71885 BECMC71886 BECMC71887 BECMC71888 BECMC71889 BECMC71900 BECMC71901 BECMC71902 BECMC71903 BECMC71904 BECMC71905 BECMC71906 BECMC85213 BECMC85214
Bench top cell sorter, CytoFLEX SRT
Zellzähler und Durchflusszytometer Durchflusszytometer
If you need an approachable instrument for sorting multiple populations to generate high quality material for downstream assays, the CytoFLEX SRT benchtop cell sorter is quick to learn and easy to operate. This extension of the CytoFLEX platform is built on the same optical features as the flow cytometer, but equally important, it continues to adhere to the principles that made multicolor applications accessible to many biomedical research, core research, pharma and biotechs.
  • Exquisite sensitivity for multicolor applications, up to 15 fluorescent colors to