Extraction unit, MSE29, Behrotest®
Lieferant: BEHR
B00217680
BEHRB00217680EA 263 EUR
BEHRB00217680
Extraction unit, MSE29, Behrotest®
Extraktoren Flüssig-Flüssig-Extraktoren
Behrotest funnel separator is used for the determination of PAHs from water.
The design of Behrotest® funnel separator facilitates simple, clean separation of the phases or the emulsion. The entire organic phase can easily be transferred in the movable separator funnel top for further processing.
The Behrotest® funnel separator is used for the isolation of lipophilic substances from water using liquid phase extraction. The various functional groups of the system can be separated. It also makes easy to clean the individual functional groups.
Standards: Conforms with DIN 38407 F18.