Incubator thermometer, DURAC® Plus™, SP Bel-Art
Lieferant: Bel-Art Products, a Part of SP
B606000200
BELAB606000200EA 205 EUR
BELAB606000200
Incubator thermometer, DURAC® Plus™, SP Bel-Art
Thermometer Analoge Thermometer
This mercury-free thermometer is designed to verify sample temperatures in incubators, and cover ranges from 10 to 50 °C.
  • Accurate temperature reading without sample contamination is achieved by using the filled bottle (included), which mimics the sample
  • Features permanently fused markings and lead-free glass
  • Insert thermometer into the bottle and place near the samples for best results
Each instrument is individually calibrated and certified against equipment whose calibration is traceable to the National Institute of Standards and Technology.
Standards: Traceable to NIST, individually serialised.
Lieferumfang: Includes multi-language (English, French, German, Italian, Portuguese, and Spanish) statement of accuracy and traceability.