Wash bottles, narrow neck, SP Bel-Art
Lieferant: Bel-Art Products, a Part of SP
F116130250 F116150250 F116140250 F116130500 F116140500 F116150500 F116170250 F116170500 F116180008 F116180016 F116131000 F116141000 F116180032 F116151000 F116171000
BELAF116130250EA 89.8 EUR
BELAF116130250 BELAF116150250 BELAF116140250 BELAF116130500 BELAF116140500 BELAF116150500 BELAF116170250 BELAF116170500 BELAF116180008 BELAF116180016 BELAF116131000 BELAF116141000 BELAF116180032 BELAF116151000 BELAF116171000
Wash bottles, narrow neck, SP Bel-Art
Flaschen Spritzflaschen
LDPE, with PP cap.
  • Available with spray caps in five colours
  • Instantaneous recognition of contents by colored cap
  • Easy-to-squeeze