Trigger sprayers, SP Bel-Art
Lieferant: Bel-Art Products, a Part of SP
F116200050
BELAF116200050EA 98 EUR
BELAF116200050
Trigger sprayers, SP Bel-Art
Verschlüsse Spritzverschlüsse
PP, durable, chemical resistant.
  • Stainless steel, spring-action pump in the trigger sprayer will not corrode with repeated exposure to 10% bleach disinfecting solution
  • Sprayer without adapter fits most bottles with a 28 mm opening
  • Adapter plus trigger sprayer adds approx. 140 mm height to a wash bottle
Trigger sprayers with adapters fit any Scienceware® bottle with an easy to fill, 53 mm wide closure and offer convenient, adjustable spray dispensing of a wide range of chemicals including bleach, ethanol, machine oil and water. Simple to install sprayer is perfect for GHS Labelled safety-vented wash bottles (series 12416), as well as OSHA compliant 16 and 32 oz. Scienceware® non-vented, wide-mouth wash bottles