Multipurpose trays, with lid, SP Bel-Art
Lieferant: Bel-Art Products, a Part of SP
F162300000
BELAF162300000EA 107 EUR
BELAF162300000
Multipurpose trays, with lid, SP Bel-Art
Schalen/Schüsseln/Töpfe Laborschalen und Laborwannen
PP.
  • Chemical and corrosion resistant
  • Flexible and strong
  • Stackable to conserve space
Multipurpose trays with snap on lid is ideal for safely storing small items and holding instruments while autoclaving. Rounded and smooth inside corners allow easy cleaning.